accessibility menu, dialog, popup

UserName

CH3HNNH2CH3HCl

CAS RN 60481-36-9

IUPAC Name: (3,5-dimethylphenyl)hydrazine hydrochloride
Synonyms: (3,5-dimethylphenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 172.66
Molecular Formula: C8H13ClN2
InChi Key: RSBQANBNDXZFIY-UHFFFAOYSA-N
SMILES: Cl.CC1=CC(NN)=CC(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)