accessibility menu, dialog, popup

UserName

H3CCH3OOCH3OOH3C

CAS RN 6065-63-0

IUPAC Name: 1,3-diethyl 2,2-dipropylpropanedioate
Synonyms: 1,3-diethyl 2,2-dipropylpropanedioate
Molecular Weight (g/mol): 244.33
Molecular Formula: C13H24O4
InChi Key: NNDOHYGFLASMFR-UHFFFAOYSA-N
SMILES: CCCC(CCC)(C(=O)OCC)C(=O)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)