missing translation for 'onlineSavingsMsg'
Learn More

Diethyl di-n-propylmalonate, 97%, Thermo Scientific™

Product Code. 11446047
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446047 100 g 100g
11436047 25 g 25g
11456047 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446047 Supplier Thermo Scientific Alfa Aesar Supplier No. B25062.22
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6065-63-0
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.331
MDL Number MFCD00026840
InChI Key NNDOHYGFLASMFR-UHFFFAOYSA-N
PubChem CID 80168
IUPAC Name diethyl 2,2-dipropylpropanedioate
SMILES CCCC(CCC)(C(=O)OCC)C(=O)OCC

Specifications

Quantity 100 g
Synonym diethyl dipropylmalonate, diethyl 2,2-dipropylmalonate, diethyl-dipropylmalonate, diethyldipropylmalonate, diethyl di-n-propylmalonate, 1,3-diethyl 2,2-dipropylpropanedioate, propanedioic acid, dipropyl-, diethyl ester, malonic acid, dipropyl-, diethyl ester, acmc-209mla, diethyl2,2-dipropylmalonate
Formula Weight 244.33
Percent Purity 97%
Chemical Name or Material Diethyl di-n-propylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.