accessibility menu, dialog, popup

UserName

CAS RN 6065-82-3

H3COOH3COOCH3

CAS RN 6065-82-3

IUPAC Name: ethyl 2,2-diethoxyacetate
Synonyms: ethyl 2,2-diethoxyacetate
Molecular Weight (g/mol): 176.21
Molecular Formula: C8H16O4
InChi Key: XCLBIKIQSCTANZ-UHFFFAOYSA-N
SMILES: CCOC(OCC)C(=O)OCC
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)