missing translation for 'onlineSavingsMsg'
Learn More

Ethyl diethoxyacetate, 98%

Product Code. 10367280 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10367280

Brand: Thermo Scientific Acros 156071000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6065-82-3 | C8H16O4 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6065-82-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00009231
InChI Key XCLBIKIQSCTANZ-UHFFFAOYSA-N
Synonym ethyl diethoxyacetate, acetic acid, diethoxy-, ethyl ester, diethoxyacetic acid ethyl ester, ethyldiethoxyacetate, acetic acid, 2,2-diethoxy-, ethyl ester, acetic acid,diethoxy,ethyl ester, acmc-209mlb, ethyl bis ethyloxy acetate, ethyl glyoxylate diethyl acetal
PubChem CID 80169
IUPAC Name ethyl 2,2-diethoxyacetate
SMILES CCOC(C(=O)OCC)OCC

Specifications

Density 0.9890g/mL
Boiling Point 198°C to 200°C (761.0 mmHg)
Flash Point 85°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2CHCO2C2H5
Refractive Index 1.41
Quantity 100 g
Beilstein 03, 601
Fieser 06,253
Specific Gravity 0.989
Formula Weight 176.21
Percent Purity 98%
Chemical Name or Material Ethyl diethoxyacetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.