accessibility menu, dialog, popup

UserName

CAS RN 607-28-3

IUPAC Name: 3-(hydroxyamino)-2H-indol-2-one
Synonyms: 3-(hydroxyamino)indol-2-one
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: LNMAXZZQNSPQSR-UHFFFAOYSA-N
SMILES: ONC1=C2C=CC=CC2=NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results