missing translation for 'onlineSavingsMsg'
Learn More

Isatin-3-oxime, 97%, Thermo Scientific™

Product Code. 11449747
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449747 100 g 100g
11429747 5 g 5g
11439747 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449747 Supplier Thermo Scientific Alfa Aesar Supplier No. B22732.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-28-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00014568
InChI Key LNMAXZZQNSPQSR-UHFFFAOYSA-N
Synonym isatin-3-oxime, 3-hydroxyimino indolin-2-one, 1h-indole-2,3-dione 3-oxime, 3-oximeindole-2,3-dione, isatin beta-oxime, 1h-indole-2,3-dione, 3-oxime, 3-hydroxyiminoindolin-2-one, indole-2,3-dione, 3-oxime, 3e-1h-indole-2,3-dione 3-oxime, 3-hydroxyamino indol-2-one
PubChem CID 5351629
IUPAC Name 3-(hydroxyamino)indol-2-one
SMILES ON=C1C(=O)NC2=C1C=CC=C2

Specifications

Melting Point ∼228°C (decomposition)
Quantity 100 g
Beilstein 136198
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Isatin-3-oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.