CAS RN 60811-18-9
IUPAC Name:
4-bromo-1-chloro-2-fluorobenzene
Synonyms:
4-bromo-1-chloro-2-fluorobenzene
Molecular Weight (g/mol):
209.44
Molecular Formula:
C6H3BrClF
InChi Key:
AGYWDGVTLKNTBS-UHFFFAOYSA-N
SMILES:
FC1=C(Cl)C=CC(Br)=C1
1
–
1
of
1
results
1
4-Bromo-1-chloro-2-fluorobenzene, 98%
CAS: 60811-18-9 Molecular Formula: C6H3BrClF Molecular Weight (g/mol): 209.44 MDL Number: MFCD00042519 InChI Key: AGYWDGVTLKNTBS-UHFFFAOYSA-N Synonym: 1-bromo-4-chloro-3-fluorobenzene,3-fluoro-4-chlorobromobenzene,2-fluoro-4-bromochlorobenzene,2-chloro-5-bromofluorobenzene,4-chloro-3-fluorobromobenzene,5-bromo-2-chlorofluorobenzene,4-bromo-1-chloro-2-fluoro-benzene,4-bromo-2-fluorochlorobenzene,benzene, 4-bromo-1-chloro-2-fluoro,3-fluoro-4-chloro-bromobenzene PubChem CID: 2724690 IUPAC Name: 4-bromo-1-chloro-2-fluorobenzene SMILES: FC1=C(Cl)C=CC(Br)=C1