CAS RN 60876-70-2
IUPAC Name:
1-bromo-4-(tert-butoxy)benzene
Synonyms:
1-bromo-4-(tert-butoxy)benzene
Molecular Weight (g/mol):
229.12
Molecular Formula:
C10H13BrO
InChi Key:
QIWQHUCUWNGYDZ-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals
CAS: 60876-70-2 Molecular Formula: C10H13BrO Molecular Weight (g/mol): 229.117 MDL Number: MFCD00792676 InChI Key: QIWQHUCUWNGYDZ-UHFFFAOYSA-N Synonym: 1-bromo-4-tert-butoxy benzene,1-bromo-4-tert-butoxybenzene,4-tert-butoxybromobenzene,4-bromophenyl tert-butyl ether,1-bromo-4-tert-butoxy-benzene,benzene, 1-bromo-4-1,1-dimethylethoxy,tert-butyl-4-bromophenyl ether,1-bromo-4-2-methylpropan-2-yl oxy benzene,1-tert-butoxy-4-bromobenzene,bromobutoxybenzene PubChem CID: 2763959 IUPAC Name: 1-bromo-4-[(2-methylpropan-2-yl)oxy]benzene SMILES: CC(C)(C)OC1=CC=C(C=C1)Br