accessibility menu, dialog, popup

UserName

H2NOCl

CAS RN 609-66-5

IUPAC Name: 2-chlorobenzamide
Synonyms: 2-chlorobenzamide
Molecular Weight (g/mol): 155.58
Molecular Formula: C7H6ClNO
InChi Key: RBGDLYUEXLWQBZ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results