accessibility menu, dialog, popup

UserName

N(R)N(R)O(S)(R)NNOHNH2HOOPHOHOO

CAS RN 61-19-8

IUPAC Name: {[5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: adenosine monophosphate
Molecular Weight (g/mol): 347.22
Molecular Formula: C10H14N5O7P
InChi Key: UDMBCSSLTHHNCD-UHFFFAOYNA-N
SMILES: NC1=C2N=CN(C3OC(COP(O)(O)=O)C(O)C3O)C2=NC=N1
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)