missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-Monophosphate, TRC

Product Code. 30138739
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138739 25 g 25g
30135275 50 g 50g
30008017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138739 Supplier Toronto Research Chemicals Supplier No. TRCA28178025G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-19-8
Molecular Formula C10 H14 N5 O7 P
Synonym 5'-Adenylic acid∣5'-AMP∣AMP∣AMP (nucleotide)∣Adenosine 5'-(dihydrogen phosphate)∣Adenosine 5'-monophosphate∣Adenosine 5'-phosphate∣Adenosine 5'-phosphoric acid∣Adenosine monophosphate∣Adenosine phosphate∣Adenosine-5-monophosphoric acid∣Adenosine-5'-monophosphoric acid∣Adenovite∣Adenylic acid∣Cardiomone∣Lycedan∣My-B-Den∣NSC 20264∣Phosaden∣Phosphaden∣Phosphentaside

Specifications

Format Neat
Molecular Weight (g/mol) 347.22
Product Type API
Shipping Condition Room Temperature
Purity >95% (HPLC)
Quantity 25 g
Chemical Name or Material 5'-Adenylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.