accessibility menu, dialog, popup

UserName

CAS RN 610-78-6

OHClNOO

CAS RN 610-78-6

IUPAC Name: 4-chloro-3-nitrophenol
Synonyms: 4-chloro-3-nitrophenol
Molecular Weight (g/mol): 173.55
Molecular Formula: C6H4ClNO3
InChi Key: JUIKCULGDIZNDI-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)