accessibility menu, dialog, popup

UserName

OOHOHOOHOHOOHOHOOHOHH2OKKKK

CAS RN 6100-19-2

IUPAC Name: bis(2,3-dihydroxybutanedioic acid) hydrate tetrapotassium
Synonyms: bis((.+-.)-tartaric acid) hydrate tetrapotassium
Molecular Weight (g/mol): 474.58
Molecular Formula: C8H14K4O13
InChi Key: DFVWEDQSAWCYPI-UHFFFAOYNA-N
SMILES: O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results