missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%

Product Code. 11935061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11935061 500 g 500g
11925061 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11935061 Supplier Thermo Scientific Acros Supplier No. 427995000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-19-2
Molecular Formula C8H14K4O13
Molecular Weight (g/mol) 474.58
MDL Number MFCD00013065
InChI Key DFVWEDQSAWCYPI-UHFFFAOYNA-N
Synonym potassium tartrate, dipotassium tartrate, argol, dipotassium 2,3-dihydroxybutanedioate, potassium tatrate, potassium tartarate, ksc492e6f
PubChem CID 6100505
IUPAC Name bis(2,3-dihydroxybutanedioic acid) hydrate tetrapotassium
SMILES O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O

Specifications

Quantity 500 g
Packaging Plastic bottle
Assay Percent Range 97%
Merck Index 15, 7796
Solubility Information Solubility in water: 226g/L (20°C).
Formula Weight 226.27
Percent Purity 97%
Chemical Name or Material L-(+)-Tartaric acid dipotassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.