accessibility menu, dialog, popup

UserName

OSOOClCl

CAS RN 610277-84-4

IUPAC Name: 4-(2-chlorophenoxy)benzene-1-sulfonyl chloride
Synonyms: 4-(2-chlorophenoxy)benzenesulfonyl chloride
Molecular Weight (g/mol): 303.15
Molecular Formula: C12H8Cl2O3S
InChi Key: ILZBGLOVCXTRDS-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1OC1=CC=C(C=C1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results