accessibility menu, dialog, popup

UserName

NaCH3ONHCCH3NH3CSOOOCH3NCH3SOOO

CAS RN 6104-58-1

IUPAC Name: sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}-2-methylphenyl)methylium
Synonyms: sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}-2-methylphenyl)methylium
Molecular Weight (g/mol): 854.02
Molecular Formula: C47H48N3NaO7S2
InChi Key: CMGWKYXJIYYESN-UHFFFAOYSA-M
SMILES: [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (14)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
Color 
  • (4)
  • (3)
  • (1)
  • (1)
Grade 
  • (3)
  • (1)
  • (4)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (14)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)