missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Blue G (Technical Grade), TRC

Product Code. 30335995
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335995 Supplier Toronto Research Chemicals Supplier No. TRCB6774031G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brilliant Blue G
CAS 6104-58-1
Molecular Formula C47 H49 N3 O7 S2 . Na
Quantity 1 g
Synonym Brilliant Indocyanine G, C.I. Acid Blue 90, C.I. Acid Blue 90, Monosodium Salt, Acid Blue 90, Acid Blue G 4061, Acid Blue PG, Acid Bright Blue G, Acid Brilliant Blue G, Acid Brilliant Cyanine G, Acidine Sky Blue G, Amacid Brilliant Cyanine G, Anadurm Cyanine A-G, BBG, Benzyl Cyanine G, Biosafe Coomassie Stain, Brilliant Acid Blue G, Brilliant Acid Blue GI, Brilliant Acid Blue J, Brilliant Acid Cyanine G, Brilliant Blue G 250, Brilliant Blue J, Brilliant Indocyanine GA-CF, Bucacid Brilliant Indocyanine G, C.I. 42655, CBB-G 250, Colocid Brilliant Blue EG, Coomassie Blue G, Coomassie Blue G 250, Coomassie Brilliant Blue G, Coomassie Brilliant Blue G 250, Coomassie G 250, Cyanine G, Daiwa Acid Blue 300, Derma Cyanine G, Derma Cyanine GN 360, Dycosweak Acid Brilliant Blue G, Eriosin Brilliant
InChI Formula InChI=1S/C47H49N3O7S2.Na/c1-6-49(31-35-11-9-13-43(29-35)58(51,52)53)40-21-25-45(33(4)27-40)47(37-15-17-38(18-16-37)48-39-19-23-42(24-20-39)57-8-3)46-26-22-41(28-34(46)5)50(7-2)32-36-12-10-14-44(30-36)59(54,55)56;/h9-30H,6-8,31-32H2,1-5H3,(H2,51,52,53,54,55,56);/q;+1
SMILES [Na+].CCOc1ccc(Nc2ccc(cc2)\C(=C\3/C=C\C(=[N+](\CC)/Cc4cccc(c4)S(=O)(=O)[O-])\C=C3C)\c5ccc(cc5C)N(CC)Cc6cccc(c6)S(=O)(=O)O)cc1
IUPAC Name sodium;3-[[(E)-[(4Z)-4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfophenyl)methyl]amino]-2-methyl-phenyl]methylene]-3-methyl-cyclohexa-2,5-dien-1-ylidene]-ethyl-ammonio]methyl]benzenesulfonate
Molecular Weight (g/mol) 855.03
Formula Weight 854.2904
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.