accessibility menu, dialog, popup

UserName

CAS RN 611-19-8

ClCl

CAS RN 611-19-8

IUPAC Name: 1-chloro-2-(chloromethyl)benzene
Synonyms: 1-chloro-2-(chloromethyl)benzene
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: BASMANVIUSSIIM-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (2)