accessibility menu, dialog, popup

UserName

BrBrNH2HNS

CAS RN 61296-22-8

IUPAC Name: 5-bromo-2-imino-2,3-dihydro-1,3-thiazol-3-ium bromide
Synonyms: 5-bromo-2-imino-3H-1,3-thiazol-3-ium bromide
Molecular Weight (g/mol): 259.95
Molecular Formula: C3H4Br2N2S
InChi Key: NUSVDASTCPBUIP-UHFFFAOYSA-N
SMILES: [Br-].BrC1=C[NH2+]C(=N)S1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results