missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromothiazole hydrobromide, 97%

Product Code. 15414117
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414117 25 g 25g
15494107 1 g 1g
15404117 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15414117 Supplier Thermo Scientific Alfa Aesar Supplier No. H31514.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61296-22-8 | C3H4Br2N2S | 259.947 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61296-22-8
Molecular Formula C3H4Br2N2S
Molecular Weight (g/mol) 259.947
MDL Number MFCD00012712
InChI Key NUSVDASTCPBUIP-UHFFFAOYSA-N
Synonym 2-amino-5-bromothiazole hydrobromide, 2-amino-5-bromothiazole monohydrobromide, 5-bromothiazol-2-amine hydrobromide, 5-bromo-1,3-thiazol-2-amine hydrobromide, 5-bromo-thiazol-2-ylamine hydrobromide, 2-amino-5-bromothiazole hbr, 2-amino-5-bromo-1,3-thiazole hydrobromide, 2-thiazolamine, 5-bromo-, monohydrobromide, 5-bromo-1,3-thiazole-2-ylamine, bromide, acmc-209mr7
PubChem CID 2723848
IUPAC Name 5-bromo-1,3-thiazol-2-amine;hydrobromide
SMILES C1=C(SC(=N1)N)Br.Br

Specifications

Melting Point ∽165°C (decomposition)
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 259.95
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromothiazole hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.