CAS RN 613-12-7
CAS RN 613-12-7
IUPAC Name:
2-methylanthracene
Synonyms:
2-methylanthracene
Molecular Weight (g/mol):
192.26
Molecular Formula:
C15H12
InChi Key:
GYMFBYTZOGMSQJ-UHFFFAOYSA-N
SMILES:
CC1=CC=C2C=C3C=CC=CC3=CC2=C1
1
–
2
of
2
results
2
2-Methylanthracene, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality