accessibility menu, dialog, popup

UserName

H3CNNOO

CAS RN 613-30-9

IUPAC Name: 2-methyl-6-nitroquinoline
Synonyms: 2-methyl-6-nitroquinoline
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: DXDPHHQJZWWAEH-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(C=CC2=N1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results