missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitroquinoline, 98%

Product Code. 11484363
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484363 25 g 25g
11464363 1 g 1g
11474363 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484363 Supplier Thermo Scientific Alfa Aesar Supplier No. A10945.14
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 613-30-9 | C10H8N2O2 | 188.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-30-9
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00051736
InChI Key DXDPHHQJZWWAEH-UHFFFAOYSA-N
Synonym 6-nitroquinaldine, quinoline, 2-methyl-6-nitro, maybridge1_001441, acmc-1bb88, 1-2-aminoethyl pyrrole-2,5-dione,hydrochloride
PubChem CID 69172
IUPAC Name 2-methyl-6-nitroquinoline
SMILES CC1=CC=C2C=C(C=CC2=N1)[N+]([O-])=O

Specifications

Melting Point 162°C to 166°C
Quantity 25 g
Beilstein 7915
Formula Weight 188.19
Percent Purity 98%
Chemical Name or Material 2-Methyl-6-nitroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.