accessibility menu, dialog, popup

UserName

HNOO(E/Z)

CAS RN 61348-47-8

IUPAC Name: 6-(2,5-dihydro-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
Synonyms: 6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: BXTIUXXTTJKNET-UHFFFAOYSA-N
SMILES: O=C1C=CC=CC1=C1ONC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results