missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Isoxazolyl)phenol, 98%, Thermo Scientific™

Product Code. 11401730
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401730 5 g 5g
11491720 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11401730 Supplier Thermo Scientific Alfa Aesar Supplier No. H27490.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61348-47-8
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00218653
InChI Key BXTIUXXTTJKNET-UHFFFAOYSA-N
Synonym phenol, 2-5-isoxazolyl, 2-isoxazol-5-yl phenol, 2-5-isoxazolyl phenol, 6-2h-1,2-oxazol-5-ylidene cyclohexa-2,4-dien-1-one, 2-1,2-oxazol-5-yl phenol, 2-isoxazol-5-ylphenol, dbdxtiaevfsdnn-uhfffaoysa-n, 2-5-isoxazolyl-phenol, acmc-1b562, 2-isoxazol-5-yl-phenol
PubChem CID 6848567
IUPAC Name 6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=C2C=CNO2)C(=O)C=C1

Specifications

Melting Point 183°C to 184°C
Quantity 5 g
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material 2-(5-Isoxazolyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.