accessibility menu, dialog, popup

UserName

CAS RN 613657-60-6

IUPAC Name: 3-[3-(trifluoromethyl)benzenesulfonamido]benzoic acid
Synonyms: 3-[3-(trifluoromethyl)benzenesulfonamido]benzoic acid
Molecular Weight (g/mol): 345.29
Molecular Formula: C14H10F3NO4S
InChi Key: LTXNSYUJJVHELV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(NS(=O)(=O)C2=CC=CC(=C2)C(F)(F)F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results