accessibility menu, dialog, popup

UserName

CAS RN 614-69-7

CH3NCS

CAS RN 614-69-7

IUPAC Name: 1-isothiocyanato-2-methylbenzene
Synonyms: 1-isothiocyanato-2-methylbenzene
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: JYKYYPPZLPVIBY-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1N=C=S
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)