missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl isothiocyanate, 98%

Product Code. 11499877
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499877 5 g 5g
11409887 25 g 25g
11419887 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499877 Supplier Thermo Scientific Alfa Aesar Supplier No. B22845.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 614-69-7 | C8H7NS | 149.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-69-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004802
InChI Key JYKYYPPZLPVIBY-UHFFFAOYSA-N
Synonym 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate
PubChem CID 69195
IUPAC Name 1-isothiocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=S

Specifications

Density 1.115
Boiling Point 239°C
Flash Point 107°C (224°F)
Odor Pungent
Refractive Index 1.6361
Quantity 5 g
UN Number UN2922
Beilstein 878487
Sensitivity Moisture sensitive
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material o-Tolyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.