accessibility menu, dialog, popup

UserName

CAS RN 615-35-0

IUPAC Name: N,N'-dimethylethanediamide
Synonyms: N,N'-dimethyloxalamide
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: IPZCJUOJSODZNK-UHFFFAOYSA-N
SMILES: CNC(=O)C(=O)NC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results