missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyloxamide, 98%

Product Code. 11483734
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483734 25 g 25g
11473734 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483734 Supplier Thermo Scientific Alfa Aesar Supplier No. A11333.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-35-0 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-35-0
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00025634
InChI Key IPZCJUOJSODZNK-UHFFFAOYSA-N
Synonym n,n'-dimethyloxalamide, nn'-dimethyloxamide, n1,n2-dimethyloxalamide, ethanediamide, n,n'-dimethyl, unii-573d5i9hqv, 4-oxalic acid n-methyl amide, ch3nhcoconhch3, n,n'-dimethylethanediamide, n,n/'-dimethyloxamide, acmc-1bg8g
PubChem CID 69204
IUPAC Name N,N'-dimethyloxamide
SMILES CNC(=O)C(=O)NC

Specifications

Melting Point 215°C to 217°C
Quantity 25 g
Beilstein 1753961
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material N,N'-Dimethyloxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.