accessibility menu, dialog, popup

UserName

CAS RN 6153-39-5

IUPAC Name: 5-methylbenzene-1,3-diol hydrate
Synonyms: orcinol hydrate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: NBKPNAMTHBIMLA-UHFFFAOYSA-N
SMILES: O.CC1=CC(O)=CC(O)=C1
Molecular Weight (g/mol) 
  • (6)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (6)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)