accessibility menu, dialog, popup

UserName

CH3OOO(S)O(S)(S)OH(S)OH(R)HOOHO

CAS RN 6160-80-1

IUPAC Name: (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxane-2-carboxylic acid
Synonyms: (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(4-methyl-2-oxochromen-7-yl)oxy]oxane-2-carboxylic acid
Molecular Weight (g/mol): 352.3
Molecular Formula: C16H16O9
InChi Key: ARQXEQLMMNGFDU-JHZZJYKESA-N
SMILES: CC1=CC(=O)OC2=CC(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)