missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Beta-D-Glucuronide, TRC

Product Code. 30138013
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138013 10 g 10g
17999534 500 mg 500mg
17908664 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138013 Supplier Toronto Research Chemicals Supplier No. TRCM33455010G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6160-80-1
Molecular Formula C16 H16 O9
Molecular Weight (g/mol) 352.29
Synonym β-D-Glucopyranosiduronic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl, Glucopyranosiduronic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl, β-D- (8CI), 4-Methyl-2-oxo-2H-1-benzopyran-7-yl β-D-glucopyranosiduronic acid, 4-Methylumbelliferone glucuronide, 4-Methylumbelliferone β-D-glucuronide, 4-Methylumbelliferyl glucuronide, 4-Methylumbelliferyl β-D-glucuronide, 4-Methylumbelliferyl-β-D-glucuronic acid, Hymecromon-7-O-4-β-D-glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
SMILES CC1=CC(=O)Oc2cc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)ccc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H16O9/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22/h2-5,11-14,16,18-20H,1H3,(H,21,22)/t11-,12-,13+,14-,16+/m0/s1
Formula Weight 352.0794
Percent Purity >95%
Chemical Name or Material 4-Methylumbelliferyl beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.