CAS RN 617-62-9
CAS RN 617-62-9
IUPAC Name:
2-methylpentanedioic acid
Synonyms:
α-methylglutaric acid (5-L-glutamyl)-L-glutamate ion(2-) α-methylene glutarate α-methylene glutarate, ion(2-)
Molecular Weight (g/mol):
146.14
Molecular Formula:
C6H10O4
InChi Key:
AQYCMVICBNBXNA-UHFFFAOYNA-N
SMILES:
CC(CCC(O)=O)C(O)=O
1
–
2
of
2
results
1
2-Methylglutaric acid, 98%
CAS: 617-62-9 Molecular Formula: C6H10O4 Molecular Weight (g/mol): 146.142 MDL Number: MFCD00002661 InChI Key: AQYCMVICBNBXNA-UHFFFAOYSA-N Synonym: 2-methylglutaric acid,alpha-methylglutaric acid,pentanedioic acid, 2-methyl,glutaric acid, 2-methyl,2-methyl-glutaric acid,2-methylglutarate,.alpha.-methylglutaric acid,2-methyl glutaric acid,2-methyl-pentanedioic acid,2-methylpentanedioate PubChem CID: 12046 ChEBI: CHEBI:68567 IUPAC Name: 2-methylpentanedioic acid SMILES: CC(CCC(=O)O)C(=O)O
2
2-Methylpentanedioic acid, MedChemExpress
MedChemExpress 2-Methylpentanedioic acid is a metabolite of succinic acid, a citric acid cycle intermediate.