missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpentanedioic acid, MedChemExpress

Product Code. 16413459
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16413459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16413459 Supplier Medchem Express Supplier No. HYW017524
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 2-Methylpentanedioic acid is a metabolite of succinic acid, a citric acid cycle intermediate.

2-Methylpentanedioic acid is a metabolite of succinic acid, a citric acid cycle intermediate.

Specifications

Chemical Name or Material 2-Methylpentanedioic acid
CAS 617-62-9
Color White
Purity Grade Notes Research
Molecular Formula C6H10O4
Quantity 1 g
Solubility Information DMSO : 100 mg/mL (684.28 mM; Need ultrasonic)
SMILES O=C(O)C(C)CCC(O)=O
Molecular Weight (g/mol) 146.14
Formula Weight 146.14
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.