CAS RN 61732-64-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
para-Cypermethrin, TRC
CAS: 61732-64-7 Molecular Formula: C22H19Cl2NO3 Molecular Weight (g/mol): 416.3 Synonym: 3-(2,2-Dichloroethenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid Cyano(4-phenoxyphenyl)methyl Ester IUPAC Name: [cyano-(4-phenoxyphenyl)methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate SMILES: Cl/C(Cl)=C\C1C(C(OC(C2=CC=C(OC3=CC=CC=C3)C=C2)C#N)=O)C1(C)C