accessibility menu, dialog, popup

UserName

H2NOOHOHHO

CAS RN 618-73-5

IUPAC Name: 3,4,5-trihydroxybenzamide
Synonyms: 3,4,5-trihydroxybenzamide
Molecular Weight (g/mol): 169.14
Molecular Formula: C7H7NO4
InChi Key: RBQIPEJXQPQFJX-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(O)=C(O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results