accessibility menu, dialog, popup

UserName

OHOOHOHO

CAS RN 618-83-7

IUPAC Name: 5-hydroxybenzene-1,3-dicarboxylic acid
Synonyms: 5-hydroxyisophthalic acid
Molecular Weight (g/mol): 182.13
Molecular Formula: C8H6O5
InChi Key: QNVNLUSHGRBCLO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(O)=C1)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)