missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyisophthalic Acid, TRC

Product Code. 30406553
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406553 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406553 Supplier Toronto Research Chemicals Supplier No. TRCH9434851G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Hydroxyisophthalic Acid
CAS 618-83-7
Purity Grade Notes HPLC
Molecular Formula C8 H6 O5
Quantity 1 g
Synonym 3,5-Dicarboxyphenol, 5-Hydroxy-1,3-benzenedicarboxylic Acid, NSC 302088
InChI Formula InChI=1S/C8H6O5/c9-6-2-4(7(10)11)1-5(3-6)8(12)13/h1-3,9H,(H,10,11)(H,12,13)
SMILES OC(=O)c1cc(O)cc(c1)C(=O)O
IUPAC Name 5-hydroxybenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol) 182.13
Formula Weight 182.0215
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.