accessibility menu, dialog, popup

UserName

CAS RN 618-84-8

NH2NOOOHO

CAS RN 618-84-8

IUPAC Name: 3-amino-5-nitrobenzoic acid
Synonyms: 3-amino-5-nitrobenzoic acid
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: ZNVHAQRPXAQKRU-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1)[N+]([O-])=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results