CAS RN 618913-30-7
CAS RN 618913-30-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AZD5904, MedChemExpress
MedChemExpress AZD5904 is a selective and irreversible inhibitor of human Myeloperoxidase (MPO) with an IC50 of 140 nM and has similar potency in mouse and rat.