missing translation for 'onlineSavingsMsg'
Learn More

AZD5904, MedChemExpress

Product Code. 16484418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484418 10 mg 10mg
16450737 50 mg 50mg
16423076 25 mg 25mg
16417147 5 mg 5mg
16405316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16484418 Supplier Medchem Express Supplier No. HY111341
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AZD5904 is a selective and irreversible inhibitor of human Myeloperoxidase (MPO) with an IC50 of 140 nM and has similar potency in mouse and rat.

AZD5904 is a selective and irreversible inhibitor of human Myeloperoxidase (MPO) with an IC50 of 140 nM and has similar potency in mouse and rat.

Specifications

Chemical Name or Material AZD5904
CAS 618913-30-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C10H12N4O2S
Quantity 10 mg
Solubility Information DMSO : 22.73 mg/mL (90.09 mM; Need ultrasonic)
SMILES O=C1NC(N(C[C@@H]2OCCC2)C3=C1N=CN3)=S
Molecular Weight (g/mol) 252.29
Formula Weight 252.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.