accessibility menu, dialog, popup

UserName

OHNOO

CAS RN 619-25-0

IUPAC Name: (3-nitrophenyl)methanol
Synonyms: 3-nitrobenzyl alcohol M-nitrobenzyl alcohol
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: CWNPOQFCIIFQDM-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)[N+]([O-])=O
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)