accessibility menu, dialog, popup

UserName

CH3OONOO

CAS RN 619-50-1

IUPAC Name: methyl 4-nitrobenzoate
Synonyms: methyl P-nitrobenzoate
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: YOJAHJGBFDPSDI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results