accessibility menu, dialog, popup

UserName

CAS RN 6199-67-3

H3COOCH3H3CO(R)H3COH(R)(R)HO(S)CH3(S)(R)(S)OHOCH3H3C(R)CH3O(R)H3C(E)

CAS RN 6199-67-3

IUPAC Name: (3E,6R)-6-[(1R,2R,3aS,3bS,8S,9aR,9bR,11aR)-2,8-dihydroxy-3a,6,6,9b,11a-pentamethyl-7,10-dioxo-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-6-hydroxy-2-methyl-5-oxohept-3-en-2-yl acetate
Synonyms: amarine datiscn principle B cucurbitacin b 2-sulfate datisca principle B cucurbitacine B 1,2-dihydro-α-elaterin cucurbitacin B 2-sulfate datiscacin cucurbitacin B
Molecular Weight (g/mol): 558.71
Molecular Formula: C32H46O8
InChi Key: IXQKXEUSCPEQRD-DKRGWESNSA-N
SMILES: CC(=O)OC(C)(C)\C=C\C(=O)[C@](C)(O)[C@H]1[C@H](O)C[C@@]2(C)[C@@H]3CC=C4[C@@H](C[C@H](O)C(=O)C4(C)C)[C@]3(C)C(=O)C[C@]12C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (4)