missing translation for 'onlineSavingsMsg'
Learn More

Cucurbitacin B, MedChemExpress

Product Code. 16498968
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498968 10 mg 10mg
16494827 50 mg 50mg
16491398 5 mg 5mg
16443866 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498968 Supplier Medchem Express Supplier No. HYN0416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cucurbitacin B belongs to a class of highly oxidized tetracyclic triterpenoids; could repress cancer cell progression.

Cucurbitacin B belongs to a class of highly oxidized tetracyclic triterpenoids

Specifications

Chemical Name or Material Cucurbitacin B
CAS 6199-67-3
Color White
Purity Grade Notes Research
Molecular Formula C32H46O8
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (178.99 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O[C@H](C1)[C@@]([C@@](C)(O)C(/C=C/C(OC(C)=O)(C)C)=O)([H])[C@](C2)(C)[C@]1(C)[C@]3([H])CC=C4C(C)(C)C([C@@H](O)C[C@@]4([H])[C@]3(C)C2=O)=O
Molecular Weight (g/mol) 558.7
Formula Weight 558.7
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.