CAS RN 62-54-4
CAS RN 62-54-4
IUPAC Name:
calcium diacetate
Synonyms:
calcium acetate gray acetate calcium ethanoate brown acetate acetate of lime teltozan calac procalamine calcium diacetate sorbo-calcian
Molecular Weight (g/mol):
158.17
Molecular Formula:
C4H6CaO4
InChi Key:
VSGNNIFQASZAOI-UHFFFAOYSA-L
SMILES:
[Ca++].CC([O-])=O.CC([O-])=O
1
–
2
of
2
results
2
Calcium Acetate, Extra Pure, SLR, Dried, Fisher Chemical™
CAS: 62-54-4 Molecular Formula: C4H6CaO4 Molecular Weight (g/mol): 158.17 MDL Number: 12448 InChI Key: VSGNNIFQASZAOI-UHFFFAOYSA-L IUPAC Name: calcium diacetate SMILES: [Ca++].CC([O-])=O.CC([O-])=O