missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium acetate hemihydrate, BAKER ANALYZED™, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
:
500 g
Specifications
Specifications
| CAS | 62-54-4 |
| Quantity | 500 g |
| Molecular Formula | C4H6CaO4 |
| MDL Number | MFCD00012448 |
| InChI Key | VSGNNIFQASZAOI-UHFFFAOYSA-L |
| SMILES | [Ca++].CC([O-])=O.CC([O-])=O |
| IUPAC Name | calcium diacetate |
| Molecular Weight (g/mol) | 158.17 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction