accessibility menu, dialog, popup

UserName

CH3Br

CAS RN 620-13-3

IUPAC Name: 1-(bromomethyl)-3-methylbenzene
Synonyms: 1-(bromomethyl)-3-methylbenzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: FWLWTILKTABGKQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CBr)=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)